Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloroacetic acid (DCA) 3% in toluene deblocking reagent

Supplier: MilliporeSigma
Synonyms: DCA

BI0832-2505 BI0832-NP20 BI0832-4000 BI0832-NP20 BI0832-2505 BI0832-4000
EM-BI0832-2505CS 1839 USD InStock
CABI0832-NP20 EM-BI0832-2505 CABI0832-4000 CABI0832-2505 EM-BI0832-4000 EM-BI0832-NP20
Dichloroacetic acid (DCA) 3% in toluene deblocking reagent
Dichloroacetic acid (DCA)
These deblocking reagents are mainly involved in DNA synthesis.

  • Infrared spectrum conforms to the standard
  • Corrosive and toxic
  • Clear liquid form


Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
MDL Number: MFCD00004223
CAS Number: 79-43-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty