Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloroacetic acid (DCA) 3% in dichloromethane deblocking reagent

Supplier: MilliporeSigma
Synonyms: DCA

Novabiochem®
BI0833-4000 BI0833-4000
EM-BI0833-4000CS 4000 USD InStock
CA-BI0833-4000 EM-BI0833-4000
Dichloroacetic acid (DCA) 3% in dichloromethane deblocking reagent
Dichloroacetic acid (DCA)
These deblocking reagents are used for biosynthesis. Contains 3% dichloroacetic acid.

  • Clear liquid
  • Corrosive and toxic


Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
MDL Number: MFCD00004223
CAS Number: 79-43-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty