Would you like to log in again?
Supelco® pH Buffer Solutions, MilliporeSigma
Supplier: MilliporeSigma
BX16281-500ML EM0186-2 EM01822-500ML BX16411-500ML BX16341-500ML BX16341-500ML EM0186-2 EM01862-500ML BX1633-1 BX1627-1 BX1642-1 BX1632-3 BX16321-500ML BX16321-4L EM0180-2 EM01802-500ML BX16331-20L BX1626-1 BX16521-500ML BX16521-500ML BX16331-4L BX16351-500ML BX16281-4L BX16321-500ML BX16571-500ML BX1627-1 BX16281-20L BX16561-500ML BX16281-4L BX1633-1 BX1641-1 BX16411-500ML BX16331-500ML EM01822-500ML BX16551-500ML BX1633-3 BX1634-1 BX1641-1 BX1655-1 BX1634-1 BX1635-1 BX1632-4 BX1656-1 BX16331-4L BX16331-20L BX1657-1 BX16421-500ML BX1633-4 BX1628-3 BX16321-20L BX1636-1 BX1628-1 BX16551-500ML BX1632-1 EM0180-2 BX1657-1 BX16351-500ML EM01802-500ML BX16281-20L BX1655-1 BX1632-4 BX1636-1 BX1652-1 BX16361-500ML BX1656-1 BX16421-500ML BX1632-3 BX16281-500ML BX1658-1 BX1628-1 BX1642-1 BX1652-1 EM01862-500ML BX1633-3 BX16321-20L BX16321-500ML BX16321-4L BX1628-4 BX1633-4 BX1626-1 BX16331-500ML BX1628-4 BX16561-500ML BX1635-1 BX16361-500ML BX16571-500ML BX1628-3
EM-BX1642-1CS 176.91 USD InStock
EM-BX1642-1 CABX1642-1 CAEM01822500ML CADBX1626-1 EMBX16421500ML EM-BX1627-1 EMBX16521500ML EMBX16321500ML CABX16321500ML CABX16321-20L CABX16521500ML EMBX16281-4L CAEM0186-2 CABX16421500ML CAEM0180-2 CABX16551500ML CABX16321-4L EMBX16561500ML EM-BX1652-1 EMBX16281500ML EM-EM0180-2 CABX16351500ML EM-BX1658-1 CABX1652-1 CABX1633-1 CABX1656-1 EM-BX1633-4 EM-BX1633-3 EM-BX1635-1 EMBX16411500ML EM-BX1656-1 CABX1633-3 CABX1635-1 EM-BX1633-1 EMBX16361500ML CABX16281-20L EMBX16321-20L CABX16331-4L EMBX16331500ML EM-EM0186-2 CABX16331-20L CABX1628-1 CABX1628-3 CABX16281-4L CABX1628-4 EMBX16331-4L EM-BX1641-1 CABX1641-1 EMBX16571500ML CABX16341500ML EMBX1628-1 CABX16281500ML EM-BX1628-3 EM-BX1628-4 EMBX16341500ML EMEM01802500ML EMEM01862500ML CABX16571500ML CABX1636-1 EMBX16321-4L EM-BX1632-1 EMD-BX1626-1 EM-BX1636-1 EM-BX1657-1 CABX1632-1 CABX1657-1 EM-BX1632-4 EM-BX1632-3 EM-BX1634-1 CABX1632-4 CAEM01862500ML EM-BX1655-1 CABX1634-1 CABX1655-1 CABX1632-3 EMEM01822500ML CABX16331500ML EMBX16281-20L EMBX16351500ML EMBX16331-20L CA-BX1633-4 EMBX16551500ML CABX1627-1 CAEM01802500ML CABX16561500ML CABX16411500ML CABX16361500ML
Supelco® pH Buffer Solutions, MilliporeSigma
Buffers pH Calibration Buffers
Certified reference material, traceable to NIST Standard.
  • Ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
Available as colorless or color-coded solutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty