Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Cyclohexane ≥99.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Hexahydro-BenzeneHexamethylene

CX2290-1 CX2290-1 CX2290-3 CX2290-3 CX2290-5 CX2290-30 CX2290-30 CX2290-5
EM-CX2290-30EA 8165 USD InStock
EM-CX2290-30 CA-CX2290-30 EM-CX2290-3 EM-CX2290-1 CACX2290-1 CACX2290-3 EM-CX2290-5 CACX2290-5
Cyclohexane ≥99.0%, GR ACS, Supelco®
Cyclohexane
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs


Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg
Order Now
Specification Test Results
Assay (GC) 99.0% min
Appearance Clear
Color (APHA) 10 max
Form Liquid
Infrared spectrum Conforms to standard
Residue after evaporation 0.002% max
Substances darkened by sulfuric acid To pass test
Water (H2O) 0.02% max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty