Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloroacetic acid (DCA) ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: DCA

M-Clarity™ quality level = MQ200

DX0657-4000 DX0657-4000
EM-DX0657-4000CS 10201 USD InStock
CADX0657-4000 EM-DX0657-4000
Dichloroacetic acid (DCA) ≥99%, Sigma-Aldrich®
Dichloroacetic acid (DCA)


Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: ∼–4 °C
Density: 1.563 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
EINECS: 201-207-0
UN: 1764
ADR: 8,II
Merck Index: 13,03075
Order Now
Specification Test Results
Absorbance at 310 nm Report onlyAU
Assay (titration) 99% min
Form Clear liquid
Freezing point 12°C min
Identity (IR-spectrum) Monochloroacetic acid
0.20% max Trichloroacetic acid
1.00% max Water (H2O)
500ppm max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty