Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dichloroethane ≥99.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Ethylene chloride Ethylene dichloride

DX0800-2 DX0800-3 DX0800-3 DX0800-5 DX0800-2
EM-DX0800-5EA 1885 USD InStock
CADX0800-3 EM-DX0800-5 EM-DX0800-3 EM-DX0800-2 CADX0800-2
1,2-Dichloroethane ≥99.0%, GR ACS, Supelco®
1,2-Dichloroethane
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs


Formula: ClH₂CCH₂Cl
MW: 98.96 g/mol
Boiling Pt: 83.5 °C (1013 hPa)
Melting Pt: –35.5 °C
Density: 1.254 g/cm³ (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3,II
Merck Index: 13,03831
https://us-legacy.vwr.com/stibo/web/std.lang.all/54/88/725488.jpg
Order Now
Specification Test Results
Assay (GC) 99.0% min
Appearance Clear
Color (APHA) 10 max
Form Liquid
Residue after evaporation 0.002% max
Titrable acid 0.0003meq/g max
Water (H2O) 0.03% max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty