Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dithizone ≥85.0% ACS, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: 1,5-Diphenylthiocarbazone

DX2370-3 DX2370-3
EM-DX2370-3EA 1296 USD InStock
CADX2370-3 EM-DX2370-3
Dithizone ≥85.0% ACS, Supelco®
Dithizone


Formula: C₆H₅NHNHCSN=NC₆H₅
MW: 256.33 g/mol
Boiling Pt: 406 °C (1013 hPa)
Melting Pt: 168 °C
MDL Number: MFCD00003025
CAS Number: 60-10-6
EINECS: 200-454-1
Merck Index: 13,03413
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty