Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl ether, anhydrous ≥99.0% stabilized, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: 2,2'-Oxybisethane2,2'-OxydiethaneEtherEthoxyethaneEthyl ether

EX0190-3 EX0190-9 EX0190-20 EX0190-9 EX0190-5 EX0190-4 EX0190-6 EX0190-8 EX0190-5 EX0190-8 EX0190-4 EX0190-3 EX0190-20 EX0190-6
EM-EX0190-9CS 3243 USD InStock
CAEX0190-3 CAEX0190-8 EM-EX0190-9 CAEX0190-9 CAEX0190-4 EM-EX0190-6 EM-EX0190-20 CAEX0190-5 EM-EX0190-5 EM-EX0190-8 CAEX0190-6 EM-EX0190-4 EM-EX0190-3 CAEX0190-20
Diethyl ether, anhydrous ≥99.0% stabilized, GR ACS, Supelco®
Diethyl ether
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Expiration date Exp 24 months from mfg date
Assay (GC) 99.0% min
Alcohol(C2H5OH) (limit about 0.05%) To pass test
Carbonyl (as HCHO) 0.001% max
Color (APHA) 10 max
Form Clear liquid
Peroxide (as H2O2) 1.0ppm max
Residue after evaporation 0.001% max
Titrable acid 0.0002meq/g max
Water (H2O) 0.03% max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty