Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heptane (mixture of isomers), Supelco®

Supplier: MilliporeSigma
Danger

HX0090-3 HX0090-1 HX0090-3 HX0090-1
EM-HX0090-3EA 1557 USD InStock
CAHX0090-1 EM-HX0090-3 CAHX0090-3 EM-HX0090-1
Heptane (mixture of isomers), Supelco®
Heptane (mixture of isomers)
Technical


Formula: H₃C(CH₂)₅CH₃
MW: 100.2 g/mol
Boiling Pt: 97…98 °C (1013 hPa)
Melting Pt: –90.5 °C
Density: 0.68 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/73/704373.jpg
Order Now
Specification Test Results
Boiling range 88-99°C
Color (APHA) 20 max
Form Clear liquid
Infrared spectrum Conforms to standard

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty