Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexane (mixture of isomers) ≥98%, Practical grade, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: HexanesIsohexane

HX0300-1 HX0300-3 HX0300-1 HX0300-3
EM-HX0300-1CS 1946 USD InStock
EM-HX0300-1 EM-HX0300-3 CAHX0300-3 CAHX0300-1
Hexane (mixture of isomers) ≥98%, Practical grade, Supelco®
Hexane (mixture of isomers)
Practical


Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/68/16640568.jpg
Order Now
Specification Test Results
Assay (n-hexane) Report
Assay (total hexane isomers + methylcyclopentane) 98% min
Color (APHA) 20 max
Form Clear liquid
Infrared spectrum Conforms

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty