Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 34 - 37%, OmniTrace® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Muriatic acid

M-Clarity™ quality level = MQ100

HX0607-4 HX0607-2 HX0607-2 HX0607-1 HX0607-1
EM-HX0607-2EA 321 USD InStock
EM-HX0607-2 EM-HX0607-1 CAHX0607-1 EM-HX0607-4 CAHX0607-2
Hydrochloric acid 34 - 37%, OmniTrace® for trace metal analysis, Supelco®
Hydrochloric acid
For elemental metal analysis via atomic absorption and ICP instruments. Actual Lot Analysis on label. Color-coded label to match acid cap.


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Aluminum (Al) 1ppb max
Antimony (Sb) 0.5ppb max
Arsenic (As) 0.5ppb max
Assay 34-37%
Barium (Ba) 0.1ppb max
Beryllium (Be) 0.1ppb max
Bismuth (Bi) 0.1ppb max
Boron (B) 1ppb max
Bromide (Br) 10ppm max
Cadmium (Cd) 0.1ppb max
Calcium (Ca) 1ppb max
Cerium (Ce) 0.1ppb max
Cesium (Cs) 0.1ppb max
Chromium (Cr) 0.5ppb max
Cobalt (Co) 0.1ppb max
Color (APHA) 10 max
Copper (Cu) 0.5ppb max
Density at 25°C 1.17-1.18g/ml
Dysprosium (Dy) 0.1ppb max
Erbium (Er) 0.1ppb max
Europium (Eu) 0.1ppb max
Free chlorine (as Cl) 0.5ppm max
Gadolinium (Gd) 0.1ppb max
Gallium (Ga) 0.1ppb max
Gold (Au) 0.5ppb max
Hafnium (Hf) 0.1ppb max
Holmium (Ho) 0.1ppb max
Indium (In) 0.1ppb max
Iron (Fe) 1ppb max
Lanthanum (La) 0.1ppb max
Lead (Pb) 0.1ppb max
Lithium (Li) 0.1ppb max
Lutetium (Lu) 0.1ppb max
Magnesium (Mg) 0.5ppb max
Manganese (Mn) 0.1ppb max
Mercury (Hg) 0.1ppb max
Molybdenum (Mo) 0.1ppb max
Neodymium (Nd) 0.1ppb max
Nickel (Ni) 0.5ppb max
Niobium (Nb) 0.1ppb max
Potassium (K) 1ppb max
Praseodymium (Pr) 0.1ppb max
Rhenium (Re) 0.1ppb max
Rhodium (Rh) 0.1ppb max
Rubidium (Rb) 0.1ppb max
Ruthenium (Ru) 0.1ppb max
Samarium (Sm) 0.1ppb max
Scandium (Sc) 0.1ppb max
Selenium (Se) 1ppb max
Silver (Ag) 1ppb max
Sodium (Na) 1ppb max
Strontium (Sr) 0.1ppb max
Tellurium (Te) 0.1ppb max
Terbium (Tb) 0.1ppb max
Thallium (Tl) 0.1ppb max
Thorium (Th) 0.1ppb max
Thulium (Tm) 0.1ppb max
Tin (Sn) 0.5ppb max
Titanium (Ti) 0.5ppb max
Total phosphorus (P) 0.01ppm max
Total sulfur (as S) 0.3ppm max
Tungsten (W) 0.1ppb max
Uranium (U) 0.1ppb max
Vanadium (V) 0.5ppb max
Ytterbium (Yb) 0.1ppb max
Yttrium (Y) 0.1ppb max
Zinc (Zn) 1ppb max
Zirconium (Zr) 0.1ppb max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty