Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: CarbinolMethyl alcoholMethyl hydroxideMethylolWood alcohol

MX0488-6 MX0488-31 MX0488-1 MX0488-NP20 MX0488P-1 MX0488-1 MX0488-1 MX0488-36 MX0488-40 MX0488-39 MX0488-6 MX0488P-1 MX0488-NP20
CAMX0488-6CS 1496 USD InStock
CAMX0488-6 CAMX0488-1 EM-MX0488P-1 EM-MX0488-NP20 CAMX0488-NP20 CAMX0488-40 CAMX0488-36 CAMX0488P-1 CAMX0488-39 EM-MX0488-1 CATXMX0488-1P EMMX0488-31 EM-MX0488-6
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Methanol
Suitable for use in spectrophometry and HPLC and gas chromatography. Gradient controlled for absence of lipophilic interferences at 254 and 235nm. UV cutoff 203nm. Certificate of Analysis on label.


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Assay (GC) 99.9% min
Acetone 10ppm max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Gradient at 235 nm 0.001AU max
Gradient at 254 nm 0.001AU max
Identity (IR-spectrum) Conforms
Residue after evaporation 1ppm max
Titrable acid 0.3µeq/g max
Titrable base 0.2µeq/g max
UV Abs. at 203 nm 1.00AU max
UV Abs. at 205 nm 0.80AU max
UV Abs. at 210 nm 0.25AU max
UV Abs. at 220 nm 0.10AU max
UV Abs. at 230 nm 0.03AU max
UV Abs. at 250 nm 0.01AU max
UV Abs. at 260 nm 0.005AU max
UV Abs. at 300 nm 0.005AU max
UV Cut-off 203nm max
Water (H2O) 0.05% max

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty