Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish) 5% stain, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

R03040-74 R03040-74
EM-R03040-74CS 1135 USD InStock
CAR03040-74 EM-R03040-74
Eosin Y (yellowish) 5% stain, Sigma-Aldrich®
Eosin Y (yellowish)
Eosin Stain Solution 5%


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
MDL Number: MFCD00005040
CAS Number: 17372-87-1
Order Now
Specification Test Results
General composition Water soluble, aqueous solution.
Absorbance of solution Expected maximum absorbance @ 515nm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty