Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid solution ≥90.5-92.8% for Babcock milk test, Supelco®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

SX1244P-4 SX1244P-4
EM-SX1244P-4CS 1602 USD InStock
CASX1244P-4 EM-SX1244P-4
Sulfuric acid solution ≥90.5-92.8% for Babcock milk test, Supelco®
Sulfuric acid
For Babcock milk test.


Formula: H₂SO₄
MW: 98.08 g/mol
Density: 1.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Assay 90.5-92.8%
Color (APHA) 30 max
Form Clear liquid
Expiration date Exp. 24 months from manufacturing date
Specific Gravity at 20/4°C 1.820-1.830

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty