Synonyms:
Oxolane
, THF
, Cyclotetramethylene oxide
, Tetramethylene oxide
, 1,4-epoxy-Butane
, Butane alpha,delta-oxide
, Oxacyclopentane
Packed in septum-sealed bottle.
TX0284-NP20
TX0284-6
TX0284-7
TX0284-7
EM-TX0284-7CS
551
USD
InStock
CATX0284-7
CATX0284-NP20
EM-TX0284-7
EM-TX0284-6
Tetrahydrofuran, anhydrous ≥99.8%, DriSolv®, Supelco®
Tetrahydrofuran
Clear and free from particulates. Stabilized with 25ppm BHT. Suitable for use in organic synthesis and other anhydrous solvent applications. Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.