Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xylene (mixture of isomers) for histology, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Dimethylbenzene

XX0060-4 XX0060-5 XX0060-5 XX0060-20 XX0060-20
EM-XX0060-4CS 700 USD InStock
EM-XX0060-4 CAXX0060-20 EM-XX0060-5 EM-XX0060-20 CAXX0060-5
Xylene (mixture of isomers) for histology, Supelco®
Xylene (mixture of isomers)
Clearing agent for routine histology and cytology applications.


Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 29.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://us-legacy.vwr.com/stibo/web/std.lang.all/44/47/704447.jpg
Order Now
Specification Test Results
Color (APHA) 10 max
Form Liquid
Residue after evaporation 0.002% max
Water (H2O) 0.05% max
Water 0.05% max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty