Would you like to log in again?
MUP Selective Supplement, MilliporeSigma
Supplier: MilliporeSigma
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1.00045.0010 1.00045.0010
EM1.00045.0010EA 244.78 USD InStock
CA1.00045.0010 EM1.00045.0010
MUP Selective Supplement, MilliporeSigma
Media for Microbiology Additives and Supplements for Microbiology
  • Less than or equal to 70% recovery on bifidobacterium animalis subsp. lactis ATCC 27536
  • Less than or equal to 70% recovery on bifidobacterium breve ATCC 15700
  • Less than or equal to 70% recovery on bifidobacterium longum ATCC 15707
  • No growth in Lactobacillus casei ATCC 393 (WDCM 00100)
  • No growth in Lactobacillus delbrueckii subsp. bulgaricus ATCC 11842 (WDCM 00102)
  • No growth in Streptococcus salivarius subsp. thermophilus DSM 20259
In North America 1.00045.0010 replaces and is identical to 1.00045.0017
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty