Would you like to log in again?
CCDA Selective Supplement, MilliporeSigma
Supplier: MilliporeSigma
1.00071.0010
EM1.00071.0010EA 340.05 USD InStock
EM1.00071.0010
CCDA Selective Supplement, MilliporeSigma
Media for Microbiology Additives and Supplements for Microbiology
  • 0.01% recovery on Escherichia coli ATCC 25922
  • 70% minimum recovery on Campylobacter jejuni ATCC 29428
  • 50% minimum recovery on Campylobacter jejuni ATCC 33291
  • 70% minimum recovery on Campylobacter coli ATCC 33559
  • 1% maximum recovery on Saccharomyces cerevisiae ATCC 9763
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty