Would you like to log in again?
Chlorine Reagent Cl₂-2 (liquid) for Chlorine test (DPD) 0.010 - 6.00 mg/L Cl₂ free chlorine: Cl₂-1 and Cl₂-2 total chlorine: Cl₂-1, Cl₂-2, and Cl₂-3 Spectroquant®
Supplier: MilliporeSigma
Spectroquant®
1.00087.0001 1.00087.0001
EM1.00087.0001CS 11241 USD InStock
EM1.00087.0001 CA1.00087.0001
Chlorine Reagent Cl₂-2 (liquid) for Chlorine test (DPD) 0.010 - 6.00 mg/L Cl₂ free chlorine: Cl₂-1 and Cl₂-2 total chlorine: Cl₂-1, Cl₂-2, and Cl₂-3 Spectroquant®
Environmental Test Kits
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty