Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orthophosphoric acid ≥75%, EMPROVE® EXPERT, SAFC®

Supplier: MilliporeSigma
Danger
Synonyms: Phosphoric acid

1.00250.9950 1.00250.9010
EM1.00250.9950EA 23245 USD InStock
CA1.00250.9010 EM1.00250.9950
Orthophosphoric acid ≥75%, EMPROVE® EXPERT, SAFC®
Orthophosphoric acid


Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 135 °C
Melting Pt: −17 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III
Order Now
Specification Test Results
Assay (alkalimetric) 74.0- 76.0 %
Residual solvents (ICH Q3C) ... excluded by production process
Endotoxins ≤ 2.5 I.U./ml

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty