Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 32% (by alkalimetry), Extra Pure, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Muriatic acidHydrochloric acid solution

1.00313.9180 1.00313.9180 1.00313.9180 1.00313.2500 1.00313.2500 1.00313.9025 1.00313.9025
EM1.00313.9025EA 649 USD InStock
CA1.00313.2500 EM1.00313.9025 TXLEM100313CBV EM1.00313.9180 CA1.00313.9025 EM1.00313.2500 CA1.00313.9180
Hydrochloric acid 32% (by alkalimetry), Extra Pure, Supelco®
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1.16 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -40 °C
Boiling Point 83 - 90 °C
Vapor Pressure 21.3 hPa (20 °C)
Density 1.16 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty