Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid fuming 37.0 - 38.0%, EMPROVE® ESSENTIAL BP, JP, NF, Ph. Eur., SAFC®

Supplier: MilliporeSigma
Danger
Synonyms: Muriatic acidHydrochloric acid solution

1.00314.1000 1.00314.9201 1.00314.2500 1.00314.9025 1.00314.9201 1.00314.2500 1.00314.1000 1.00314.9025
EM1.00314.1000EA 74 USD InStock
CA1.00314.9201 CA1.00314.9025 EM1.00314.1000 EM1.00314.9201 EM1.00314.9025 CA1.00314.1000 CA1.00314.2500 EM1.00314.2500
Hydrochloric acid fuming 37.0 - 38.0%, EMPROVE® ESSENTIAL BP, JP, NF, Ph. Eur., SAFC®
Hydrochloric acid
Soluble in water. Corrosive and toxic.


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Arsenic (As) 0.0001% max.
Boiling Point 110°C (230°F)
Bromide and Iodide (Br, I) Pass Test
Calcium (Ca) 0.001% max.
Free Bromine or Chlorine Pass test
Free Chlorine (Cl) 0.0001% max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty