Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodine(V) oxide, granular, EMSURE® 0.5-2.5 mm for analysis, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: di-Iodine pentoxidedi-Iodine pentaoxideIodopentoxide

1.00358.0100 1.00358.0100
EM1.00358.0100CS 28654 USD InStock
CA1.00358.0100 EM1.00358.0100
Iodine(V) oxide, granular, EMSURE® 0.5-2.5 mm for analysis, Supelco®
Iodine (V) oxide


Formula: I₂O₅
MW: 333.81 g/mol
MDL Number: MFCD00011356
CAS Number: 12029-98-0
UN: 1479
ADR: 5.1,III
Order Now
Specification Test Results
Solubility (20 °C) soluble
Molar Mass 333.81 g/mol
Bulk Density 2000 kg/m3
Density 4.98 g/cm3 (20 °C)
pH 1.1 (100 g/l, H2O, 25 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty