Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-2-pyrrolidone (NMP) for peptide synthesis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: N-Methyl-alpha-pyrrolidone1-Methyl-2-pyrrolidoneNMPN-Methyl-2-pyrrolidoneN-Methylpyrrolidone1-Methyl-2-pyrrolidinoneN-MethylpyrrolidinoneN-methylpyrrolidoneN-Methyl-gamma-butyrolactamN-Methyl-2-pyrrolidinoneM-Pyrol

M-Clarity™ quality level = MQ200

1.00574.4000 1.00574.4000 1.00574.2500 1.00574.2500
EM1.00574.2500CS 2532 USD InStock
CA1.00574.4000 EM1.00574.2500 EM1.00574.4000 CA1.00574.2500
N-Methyl-2-pyrrolidone (NMP) for peptide synthesis, Supelco®
N-Methyl-2-pyrrolidone (NMP)


Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 12,06197
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty