Would you like to log in again?
Sulfate Cell Test Method: photometric 50 to 500 mg/L SO₄²⁻ Spectroquant®, MilliporeSigma
Supplier: MilliporeSigma
Spectroquant®
1.00617.0001 1.00617.0001
EM1.00617.0001CS 998 USD InStock
CA1.00617.0001 EM1.00617.0001
Sulfate Cell Test Method: photometric 50 to 500 mg/L SO₄²⁻ Spectroquant®, MilliporeSigma
Environmental Test Kits
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty