Synonyms:
2-Propanone
, DMK
, Propanone
, Dimethyl ketone
1.00658.1000
1.00658.2500
1.00658.1000
1.00658.2500
EM1.00658.1000CS
615
USD
InStock
EM1.00658.1000
CA1.00658.1000
EM1.00658.2500
CA1.00658.2500
Acetone ≥99.8%, SupraSolv® for GC-MS, distilled, Supelco®
Acetone
SupraSolv® MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results. This method is used e. g. for the analysis of dioxins and furans (PCDD/PCDF) in food and water samples or for the determination of PAH (polycyclic aromatic hydrocarbons) in food.