Would you like to log in again?
Spectroquant® AOX Test Kits, MilliporeSigma
Supplier: MilliporeSigma
Spectroquant®
1.00677.0001 1.00677.0001
EM1.00677.0001EA 685 USD InStock
CA1.00677.0001 EM1.00677.0001
Spectroquant® AOX Test Kits, MilliporeSigma
Environmental Test Kits
Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty