Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 90-100%, Supelco®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ300

1.00732.2510 1.00732.1000
EM1.00732.2510EA 210 USD InStock
EM1.00732.2510 EM1.00732.1000
Sulfuric acid 90-100%, Supelco®
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty