Synonyms:
Hexanen
, Hexane
, Hexyl hydride
1.00795.2500
1.00795.1000
1.00795.1000
1.00795.2500
EM1.00795.2500CS
1018
USD
InStock
CA1.00795.1000
EM1.00795.2500
EM1.00795.1000
CA1.00795.2500
n-Hexane ≥99.8%, SupraSolv® for GC-MS, distilled, Supelco®
n-Hexane
SupraSolv® MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results. This method is used e. g. for the analysis of dioxins and furans (PCDD/PCDF) in food and water samples or for the determination of PAH (polycyclic aromatic hydrocarbons) in food.