Synonyms:
Hydroxy methane
, Methyl hydroxide
, Methyl alcohol
, Methylol
, Carbinol
, Wood alcohol
1.00837.1000
1.00837.1000
1.00837.2500
1.00837.2500
EM1.00837.2500CS
613
USD
InStock
CA1.00837.1000
EM1.00837.2500
EM1.00837.1000
CA1.00837.2500
Methanol, SupraSolv® for GC-MS, Supelco®
Methanol
SupraSolv® MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.