Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isoamyl alcohol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: 3-Methyl-1-butanolIsopentyl alcohol3-Methylbutanoliso-Pentanol

1.00979.1000 1.00979.2500 1.00979.1000 1.00979.2500 1.00979.4000 1.00979.4000
EM1.00979.4000EA 784 USD InStock
CA1.00979.2500 EM1.00979.4000 EM1.00979.1000 EM1.00979.2500 CA1.00979.4000 CA1.00979.1000
Isoamyl alcohol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Isoamyl alcohol


Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://us-legacy.vwr.com/stibo/web/std.lang.all/15/69/2301569.jpg
Order Now
Specification Test Results
Solubility 25 g/l (20 °C)
Melting Point -117 °C
Molar Mass 88.15 g/mol
Boiling Point 131 °C (1013 hPa)
Vapor Pressure 3 hPa (20 °C)
Flash Point 45 °C
Explosion Limit 1.2 - 8 %(V)
Density 0.81 g/cm3 (20 °C)
pH 7 (25 g/l, H2O, 20 °C)
Ignition Point 335 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty