Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium, foil, EMSURE® for analysis, Width 30 mm, Thickness 0.3 mm, Supelco®

Supplier: MilliporeSigma
Synonyms: Aluminum

1.01057.0250 1.01057.0250 1.01057.1000 1.01057.1000
EM1.01057.1000CS 8116 USD InStock
EM1.01057.1000 CA1.01057.0250 CA10053-782 EM1.01057.0250
Aluminium, foil, EMSURE® for analysis, Width 30 mm, Thickness 0.3 mm, Supelco®
Aluminium


Formula: Al
MW: 26.98 g/mol
MDL Number: MFCD00134029
CAS Number: 7429-90-5
Order Now
Specification Test Results
Solubility (20 °C) (reaction)
Melting Point 660 °C
Molar Mass 26.98 g/mol
Boiling Point 2467 °C
Density 2.70 g/cm3 (20 °C)
Ignition Point 400 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty