Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium chloride, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Ammonium muriateSalmiac

1.01145.9025 1.01145.9050 1.01145.9050 1.01145.0500 1.01145.5000 1.01145.1000 1.01145.5000 1.01145.1000 1.01145.9025 1.01145.0500
EM1.01145.0500CS 882 USD InStock
CA1.01145.9050 CA1.01145.1000 EM1.01145.0500 EM1.01145.9025 EM1.01145.1000 CA1.01145.0500 EM1.01145.5000 CA1.01145.9025 EM1.01145.9050 CA10754-008
Ammonium chloride, EMSURE® for analysis, Supelco®
Ammonium chloride


Formula: NH₄Cl
MW: 53.49 g/mol
Melting Pt: 335 °C
Density: 1.5256 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/71/16640571.jpg
Order Now
Specification Test Results
Solubility 372 g/l (20 °C)
Melting Point 335 °C (sublimed)
Molar Mass 53.49 g/mol
Bulk Density 500 kg/m3
Vapor Pressure 1.3 hPa (30 °C)
Density 1.52 g/cm3 (20 °C)
pH 4.5 - 5.5 (50 g/l, H2O, 20 °C)
Ignition Point 400 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty