Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead borate, Supelco®

Supplier: MilliporeSigma
Danger

1.01189.1000 1.01189.1000
EM1.01189.1000CS 72194.99 USD InStock
EM1.01189.1000 CA1.01189.1000
Lead borate, Supelco®
Lead borate


Formula: B₂O₆Pb₃
CAS Number: 35498-15-8
UN: 2291
ADR: 6.1,III
Order Now
Specification Test Results
Solubility (25 °C) soluble
Molar Mass 328.85 g/mol
Bulk Density 3870 kg/m3
Density 11.0 g/cm3 (20 °C)
pH 8.6 (650 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty