Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tetra-Chloroauric(III) acid trihydrate ≥99.5%, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Hydrogen tetrachloroaurate (III) trihydrate tetra-Chloroauric acid trihydrate

1.01582.0005 1.01582.0001 1.01582.0001 1.01582.0005
EM1.01582.0001CS 72465 USD InStock
CA1.01582.0005 EM1.01582.0001 EM1.01582.0005 CA1.01582.0001
tetra-Chloroauric(III) acid trihydrate ≥99.5%, EMSURE® for analysis, Supelco®
tetra-Chloroauric (III) acid trihydrate


Formula: HAuCl₄·3H₂O
MW: 393.83 g/mol
MDL Number: MFCD00149904
CAS Number: 16961-25-4
UN: 3260
ADR: 8,III
Order Now
Specification Test Results
Solubility 150 g/l (20 °C)
Melting Point 30 °C
Molar Mass 393.83 g/mol
Density 3.9 g/cm3 (20 °C)
pH (H2O) strongly acid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty