Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alcian blue solution used in nuclear staining, for histology, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: C.I. 74240C.N. AnAn-6Ingrain Blue 1Alcian blue 8 GX

1.01647.0500 1.01647.0500
EM1.01647.0500CS 4560.76 USD InStock
CA1.01647.0500 EM1.01647.0500
Alcian blue solution used in nuclear staining, for histology, Sigma-Aldrich®
Alcian blue 8 GX
Used to stain acidic mucosubstances

The combination of PAS staining with Alcian blue allows (at pH 2.5) the additional identification of acidic mucosubstances (glycosaminoglycans). PAS staining is used for paraffin sections and cell smears, and upon use of the ready-to-use Alcian blue staining solution, nuclei will appear blue, acidic mucosubstances light blue and polysaccharides as well as neutral mucopolysaccharides will be stained purple. Additionally, the Alcian blue staining solution (at pH 2.5) allows the identification of acidic mucosubstances without differentiation of carboxyl and sulfate groups, upon which the nuclei are counter-stained with nuclear fast red. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.


Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
MW: 1298.86 g/mol
Density: 1.006 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010720
CAS Number: 33864-99-2
IVDLogo
Order Now
Specification Test Results
Suitability for microscopy passes test
neutral mucopolysaccharides purple
polysaccharides purple
PAS negative acidic mucosubstances light blue
Nuclei blue

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty