Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium nitrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.01729.9050 1.01729.9050 1.01729.0500 1.01729.0500
EM1.01729.0500CS 7510 USD InStock
CA1.01729.0500 CA1.01729.9050 EM1.01729.0500 EM1.01729.9050
Barium nitrate, EMSURE® for analysis, Supelco®
Barium nitrate


Formula: Ba(NO₃)₂
MW: 261.34 g/mol
Melting Pt: ∼592 °C
Density: ∼3.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1,II
Merck Index: 12,01012
Order Now
Specification Test Results
Solubility 90 g/l (20 °C)
Melting Point 592 °C
Molar Mass 261.34 g/mol
Bulk Density 1900 g/m3
Vapor Pressure (20 °C)
Density 3.2 g/cm3 (20 °C)
pH 5.2 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty