Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzene-[D₆] ≥99.6% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Benzene-[²H₆]

M-Clarity™ quality level = MQ200

1.01789.0009 1.01789.0010 1.01789.0100 1.01789.0010 1.01789.0009 1.01789.0100
EM1.01789.0010EA 214 USD InStock
EM1.01789.0010 CA1.01789.0009 EM1.01789.0100 CA1.01789.0010 CA1.01789.0100 EM1.01789.0009
Benzene-[D₆] ≥99.6% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Benzene-[D6]
MagniSolv™ deuterated solvents provide:
• Reliable results in the NMR-spectra by excellent chemical purity and highest isotopic enrichment.
• Reliable deuteration degrees.
• Low residual water content.
• Determination of water content in two ways.


Formula: C₆D₆
MW: 84.07 g/mol
Boiling Pt: 79 °C (1013 hPa)
Melting Pt: 6.7 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: –11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003010
CAS Number: 1076-43-3
EINECS: 214-061-8
UN: 1114
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty