Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium oxide, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Lime

M-Clarity™ quality level = MQ200

1.02109.9025 1.02109.9025 1.02109.1000 1.02109.1000
EM1.02109.9025EA 3674 USD InStock
EM1.02109.9025 CA1.02109.1000 CA1.02109.9025 EM1.02109.1000
Calcium oxide, Supelco®
Calcium oxide


Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850 °C (1013 hPa)
Melting Pt: 2580 °C
Density: 3.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty