Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloramine-T (sodium salt) trihydrate, Extra Pure, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Chloramine T sodium salt trihydrateSodium chloro(tosyl)amide trihydrateTosylchloramide sodium trihydrate

1.02424.1000 1.02424.9050 1.02424.9050 1.02424.5000 1.02424.5000 1.02424.1000
EM1.02424.1000CS 2104 USD InStock
EM1.02424.1000 EM1.02424.5000 CA1.02424.9050 CA1.02424.5000 EM1.02424.9050 CA1.02424.1000
Chloramine-T (sodium salt) trihydrate, Extra Pure, Supelco®
Chloramine T (sodium salt) trihydrate


Formula: CH₃C₆H₄SO₂NClNa·3H₂O
MW: 281.69 g/mol
Melting Pt: 167…170 °C
Density: 1.43 g/cm³ (20 °C)
Flash Pt: 192 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149066
CAS Number: 7080-50-4
EINECS: 204-854-7
UN: 2928
ADR: 6.1 (8),II
Merck Index: 13,02085
Order Now
Specification Test Results
Assay (iodometric) 98.0 - 103.0 %
Identity (IR-spectrum) passes test
Appearance of solution passes test
pH-value (5 %; water) 8.0 - 10.0
Matter insoluble in ethanol ≤ 2 %
ortho compounds passes test
Residual solvents (ICH Q3C) excluded by manufactoring process

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty