Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform 99.0-99.4% (by GC) stabilized, EMPROVE® ESSENTIAL, SAFC®

Supplier: MilliporeSigma
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with ethanol 1,0 - 2,0 % (v/v)

1.02431.1000 1.02431.2500 1.02431.6025 1.02431.2500
EM1.02431.1000EA 301 USD InStock
EM1.02431.1000 EM1.02431.2500 EM1.02431.6025 CA1.02431.2500
Chloroform 99.0-99.4% (by GC) stabilized, EMPROVE® ESSENTIAL, SAFC®
Chloroform
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Purity (GC) 99.0 - 99.4% (w/w)
Identity (IR) Conforms
Acidity or Alkalinity Conforms
Density (d 20/4 °C) 1.474 - 1.479
Density (d 20/20 °C) 1.479 - 1.482
Boiling Range (60 - 62 °C) ≥95% (v/v)
Chloride (Cl) Conforms
Free Chlorine (as Cl) Conforms
Ethanol (GC) 1.0 - 2.0% (v/v)
Related Substances (GC) ≤1.0% (v/v)
Bromochloromethane (GC) ≤0.01%
1,2-Dichloroethene (GC) ≤0.05%
Dichloromethane (GC) ≤0.01%
Carbon Tetrachloride (GC) ≤0.01%
1,1,2-Trichloroethene (GC) ≤0.01%
Other Residual Solvents (ICH Q3C) Excluded by Manufacturing Process
Aldehydes Conforms
Foreign Chlorine Compounds Conforms
Matter Discolored by H₂SO₄ ≤10 Hazen
Evaporation Residue ≤0.0010%
Water ≤0.10%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty