Synonyms:
Methane, trichloro-
, Trichloroform
, Trichloromethane
Stabilization:
Stabilised with ethanol 1,0 - 2,0 % (v/v)
1.02431.1000
1.02431.2500
1.02431.6025
1.02431.2500
EM1.02431.1000EA
301
USD
InStock
EM1.02431.1000
EM1.02431.2500
EM1.02431.6025
CA1.02431.2500
Chloroform 99.0-99.4% (by GC) stabilized, EMPROVE® ESSENTIAL, SAFC®
Chloroform
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).