Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform, LiChrosolv® for liquid chromatography, Supelco®

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

M-Clarity™ quality level = MQ100

1.02444.2500 1.02444.4000 1.02444.4000 1.02444.1000 1.02444.1000
EM1.02444.4000EA 3232 USD InStock
CA10159-664 EM1.02444.4000 EM1.02444.1000 EM1.02444.2500 CA1.02444.1000
Chloroform, LiChrosolv® for liquid chromatography, Supelco®
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty