Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform-[D₁] ≥99.96% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Chloroform-DTrichloromethane-D1Chloroform-[²H₁]

M-Clarity™ quality level = MQ200

1.02446.0009 1.02446.0009 1.02446.0010 1.02446.0100 1.02446.0010 1.02446.0100 1.02446.0025 1.02446.0025
EM1.02446.0100EA 451 USD InStock
EM1.02446.0100 EM1.02446.0025 CA1.02446.0010 CA1.02446.0100 EM1.02446.0009 CA1.02446.0025 CA1.02446.0009 EM1.02446.0010
Chloroform-[D₁] ≥99.96% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Chloroform-[D1]
MagniSolv™ deuterated solvents provide reliable results in the NMR-spectra with extremely low residual water, excellent chemical purity and the highest isotopic enrichment.


Formula: CDCl₃
MW: 120.38 g/mol
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64.1 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty