Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Cyclohexane, SupraSolv® for GC-ECD/FID, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Hexahydro-BenzeneHexamethylene

1.02817.1000 1.02817.1000 1.02817.2500 1.02817.2500 1.02817.4000 1.02817.4000
EM1.02817.2500CS 1469 USD InStock
EM1.02817.2500 CA1.02817.4000 CA1.02817.1000 CA1.02817.2500 EM1.02817.4000 EM1.02817.1000
Cyclohexane, SupraSolv® for GC-ECD/FID, Supelco®
Cyclohexane
SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.


Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg
Order Now
Specification Test Results
Solubility 55 mg/l (20 °C)
Melting Point 6 °C
Molar Mass 84.16 g/mol
Boiling Point 81 °C (1013 hPa)
Vapor Pressure 103 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.4264 (20 °C)
Explosion Limit 1.2 - 8.3 %(V)
Density 0.78 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 260 °C
Viscosity kinematic 1.26 mm2/s (20 °C)
Evaporation number 3.5
Hazard Class 3
Hazard Class Description Flammable liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty