Would you like to log in again?
Culture Media Bases, MilliporeSigma
Supplier: MilliporeSigma
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1.07213.2500 1.02239.0500 1.01614.5000 1.15363.0500 1.07224.2500 1.05391.0500 1.03753.9025 1.07213.9025 1.08342.2500 1.07213.1000 1.11931.1000 1.08342.2500 1.02239.0500 1.08342.1000 1.07214.1000 1.11931.9025 1.15363.0500 1.03753.0500 1.08342.1000 1.07224.2500 1.03753.0500 1.01614.1000 1.07229.1000 1.11931.1000 1.10213.1000 1.05391.0500 1.02245.0500 1.02245.0500 1.07229.1000 1.07214.1000 1.01614.5007 1.10213.1000 1.07213.9025 1.01614.1000 1.03979.0500 1.04070.0500 1.11931.9025 1.07213.1000 1.03753.9025 1.03979.0500 1.04070.0500
EM1.10213.1000EA 401.06 USD InStock
EM1.10213.1000 CA1.04070.0500 EM1.11931.9025 EM1.02239.0500 EM1.05391.0500 CA1.08342.1000 CA1.02245.0500 EM1.01614.5007 EM1.01614.1000 CA1.07213.2500 EM1.07214.1000 CA1.07229.1000 CA1.07224.2500 EM1.08342.1000 CA1.07214.1000 CA1.03753.9025 EM1.11931.1000 CA1.15363.0500 EM1.07229.1000 EM1.07224.2500 CA1.08342.2500 EM1.02245.0500 CA1.02239.0500 CA1.01614.1000 EM1.03753.0500 CA1.10213.1000 EM1.03979.0500 CA1.07213.9025 CA1.07213.1000 EM1.07213.1000 EM1.08342.2500 EM1.07213.9025 EM1.04070.0500 CA1.01614.5000 EM1.15363.0500 CA1.03979.0500 CA1.05391.0500 CA1.03753.0500 CA1.11931.9025 EM1.03753.9025 CA1.11931.1000
Culture Media Bases, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
This line of culture media base products is produced in powdered and granular form.
Product # 1.01614.5000 replaces and is identical to Product # 1.01614.5007.
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media. Media is granulated, except where otherwise noted.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty