Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde ≥37% in aqueous solution stabilized for histology acid-, calcium carbonate-free, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: MethylaldehydeFormolFormalinParaformaldehyde aqueous solution

Stabilization: Stabilized with methanol 10%
M-Clarity™ quality level = MQ400

1.03999.1000 1.03999.9025 1.03999.2500 1.03999.9025 1.03999.2500 1.03999.1000
EM1.03999.1000CS 628 USD InStock
CA1.03999.9025 EM1.03999.1000 CA1.03999.1000 EM1.03999.9025 EM1.03999.2500 CA1.03999.2500
Formaldehyde ≥37% in aqueous solution stabilized for histology acid-, calcium carbonate-free, Sigma-Aldrich®
Formaldehyde


Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 2209
ADR: 8,III
Order Now
Specification Test Results
Assay (acidimetric) 37.0 - 38.0 %
Heavy metals as Pb ≤ 0.0002 %
Fe (Iron) ≤ 0.0001 %
Methanol (GC) 8-12 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty