Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

n-Heptane, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

1.04365.6190 1.04365.9011 1.04365.1000 1.04365.6025 1.04365.9011 1.04365.2511 1.04365.6190 1.04365.2500 1.04365.6025 1.04365.1000
EM1.04365.6025EA 1408 USD InStock
EM1.04365.6025 EM1.04365.6190 EM1.04365.1000 CA1.04365.9011 EM1.04365.9011 EM1.04365.2511 EM1.04365.2500 CA1.04365.1000 CA1.04365.6025 CA1.04365.6190
n-Heptane, EMPLURA®, Supelco®
n-Heptane


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –90.6 °C
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty