Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

n-Heptane, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: DipropylmethaneEptaniHeptanHeptaneHeptanenHeptyl hydrideSkellysolve c

M-Clarity™ quality level = MQ100

1.04366.0500 1.04366.2500 1.04366.2500 1.04366.0500
EM1.04366.2500CS 3239 USD InStock
EM1.04366.2500 CA1.04366.0500 CA1.04366.2500 EM1.04366.0500
n-Heptane, Uvasol® for spectroscopy, Supelco®
n-Heptane


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –90.6 °C
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now
Specification Test Results
Purity (GC) ≥ 99.3 %
Evaporation residue ≤ 0.0002 %
Water ≤ 0.005 %
Color ≤ 10 Hazen
Acidity ≤ 0.0001 meq/g
Alkalinity ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm) ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm) ≤ 1.0 ppb
Transmission (at 200 nm) ≥ 20 %
Transmission (at 210 nm) ≥ 55 %
Transmission (at 220 nm) ≥ 80 %
Transmission (at 228 nm) ≥ 90 %
Transmission (from 245 nm) ≥ 98 %
Absorbance (at 200 nm) ≤ 0.70
Absorbance (at 210 nm) ≤ 0.26
Absorbance (at 220 nm) ≤ 0.10
Absorbance (at 228 nm) ≤ 0.05
Absorbance (from 245 nm) ≤ 0.01

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty