Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) iodide, red ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Mercury diiodideMercuric iodide

M-Clarity™ quality level = MQ300

1.04428.0050 1.04428.0250 1.04428.0050 1.04428.0250
EM1.04428.0250CS 67133 USD InStock
CA1.04428.0250 CA1.04428.0050 EM1.04428.0250 EM1.04428.0050
Mercury(II) iodide, red ACS, Reag. Ph. Eur. for analysis, Supelco®
Mercury (II) iodide


Formula: HgI₂
MW: 454.4 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 259 °C
Density: 6.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011044
CAS Number: 7774-29-0
EINECS: 231-873-8
UN: 1638
ADR: 6.1,II
Merck Index: 12,05932
Order Now
Specification Test Results
Assay (argentometric, dried sust.) ≥ 99.0 %
Solubility in Potassium iodide passes test
Soluble mercury salts (as Hg) ≤ 0.05 %
Foreign heavy metals (as Pb) ≤ 0.001 %
Fe (Iron) ≤ 0.001 %
Mercury (I) (as Hg) ≤ 0.1 %
Substances not reducing by formic acid ≤ 0.02 %
Corresponds to ACS,Reag. Ph Eur

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty